Lubricant, anti-friction coating, Molykote® D-321 R
N° de catalogue GREF636264F
Fournisseur: GREEFF RW SILICONES LUBRICAN
Spécifications
- Description:Molykote® D-321 R, aerosol
- Pk:400 mL
Spécifications
À propos de cet article
Air-curing dry film lubricant in solvent.
- Air drying
- Avoidance of stick-slip
- High aging resistance
Recommended dry film thickness: 5 to 20 µm
Service temperature range from –180 to +260 °C.