Order Entry
Switzerland
ContactUsLinkComponent
Ion selective electrode, surfactant, Orion™
Ion selective electrode, surfactant, Orion™
  662-0821
Ion selective electrode, surfactant, Orion™
  662-0821

 

  • Concentration:
    Endpoint indicator
  • Connection:
    BNC, 1 m cable
  • Type:
    9342BN
  • Description:
    Surfactant plastic membrane half-cell, 9342BN
  • Pk:
    1
  • Temp.:
    0...+40 °C

 

 

This surfactant half cell is a potentiometric sensor for end point detection in titrations of anionic or cationic surfactants in a solution. A potentiometric sensor, it is a surfactant-detecting electrode in anionic or cationic titration which is specifically designed for use on the Orion 960 Autochemistry system.

Can be used on any ISE or mV meter with a BNC connection. Must be used with the double junction reference electrode.

Reference electrode