Redox/ORP/temperature electrodes, Orion™
Anbieter: Thermo Orion
Zertifikate
Über diesen Artikel
These ORP electrodes combine a platinum redox sensing electrode and a silver/silver chloride reference electrode in one body.
- Efficiently measure ORP and temperature simultaneously without a separate reference electrode in a single instance
- Available in epoxy triode low maintenance and epoxy refillable triode construction
- ORP Triode electrodes have a built-in thermistor for measuring temperature
The 9180BNMD and 9179BNMD ORP/ATC Triode electrodes have a temperature connector that is compatible with the Star Series meters.
The 9180BN and 9179BN ORP/ATC Triode electrode have a temperature connector that is compatible with the A+ Series meters.
Note: Not designed for amperometric or constant current titrations, such as Karl Fischer analysis.
Frequently Bought Together