ContactUsLinkComponent
Universal power adapter for Orion™ Versa Star™ meters
Universal power adapter for Orion™ Versa Star™ meters
Catalog # ORIOSTARA-PWR
Supplier:  Thermo Orion
undefined
Universal power adapter for Orion™ Versa Star™ meters
Catalog # ORIOSTARA-PWR
Supplier:  Thermo Orion
Specifications
  • Description:
    Universal power adapter for Orion™ Versa Star™ meters, with US, EU, UK and China plug, 90 - 260 VAC, 50/60 Hz
  • Pk:
    1
Specifications
About this item
Spare or replacement power adapter for Thermo Scientific™ Orion™ Versa Star meters.