ContactUsLinkComponent
Face seal cleaning wipes for respirators, 105
Face seal cleaning wipes for respirators, 105
Catalog # 111-1952
Supplier:  3M
undefined
Face seal cleaning wipes for respirators, 105
Catalog # 111-1952
Supplier:  3M
Specifications
  • Description:
    Face seal cleaning wipes for respirators, 105, presaturated
  • Pk:
    40
Specifications
About this item
The 105 face seal cleaner is an individually wrapped wipe for cleaning and freshening the face piece without damaging it.
  • Antiseptic
  • Will not dry out even after long periods of storage in warm conditions
  • Complies with current regulations requiring cleaning of non-disposable respiratory equipment after use
Packaging: Individually foil wrapped.