Order Entry
Switzerland
ContactUsLinkComponent
Clostridium histolyticum Collagenase type IV (from Clostridium histolyticum), MP Biomedicals
Clostridium histolyticum Collagenase type IV (from Clostridium histolyticum), MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Clostridium histolyticum Collagenase type IV (from Clostridium histolyticum), MP Biomedicals
Supplier:  MP Biomedicals
Certificates

Specifications

  • CAS No.:
    9001-12-1
  • Source:
    Clostridium histolyticum
  • Species:
    Clostridium histolyticum
  • Storage conditions:
    2...8 °C, desiccate
  • Form:
    Brown powder
  • Enzyme name:
    Collagenase type IV
  • Enzyme activity:
    >160 u/mg
  • Enzyme synonyms:
    Clostridiopeptidase A

Specifications

Related Information

Frequently Bought Together