Doxorubicin hydrochloride
Catalog # ICNA0215910110
Supplier: MP Biomedicals
CAS Number:
Specifications
- VE:10 mg
- Seq:2275
Specifications
Specification Test Results
- Form:Reddish Orange Powder
- Storage:Store at Room Temperature (15 - 30 °C)
Specification Test Results
About this item
:
:579.98 g/mol
CAS Number:25316-40-9
Formula:C₂₇H₂₉NO₁₁·HCl
MDL Number:MFCD00077757