


Xylene (mixture of isomers), clear, colourless liquid for histology
Catalog # ICNA0215869204
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:1 KIT
- Seq:0075
Specifications
Specification Test Results
- Identity Test:Passes
- FTIR:Conforms to Standard
- Water:≤0.05%
Specification Test Results
About this item
:
:106.17 g/mol
Boiling Pt:137…140 °C (lit.)
Melting Pt:–47.9…+13.3 °C (lit.)
Density:0.86 g/mL (25 °C) (lit.)
Storage Temperature:Ambient
UN:1307
ADR:3,III
:GHS02
:GHS07
:GHS08
CAS Number:1330-20-7
Formula:C₆H₄(CH₃)₂
MDL Number:MFCD00077264