Order Entry
Switzerland
ContactUsLinkComponent
Light green SF yellowish, purple powder C.I. 42095
Light green SF yellowish, purple powder C.I. 42095
Catalog # ICNA0215264125
Supplier:  MP Biomedicals
CAS Number:  
Light green SF yellowish, purple powder C.I. 42095
Catalog # ICNA0215264125
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Identity Test :
    Passes
  • UV/VIS: λmax (H₂O):
    630 ± 5 nm

Specification Test Results

About this item

 :
 :792.86 g/mol
Storage Temperature:Ambient
CAS Number:5141-20-8
Formula:C₃₇H₃₄N₂Na₂O₉S₃
MDL Number:MFCD00012121