Malate dehrdrogenase, MP Biomedicals
Soluble in distilled water or dilute buffer; dissolves readily at 5 mg/ml in 0,1 M potassium phosphate pH 7,4.
- GOT ≤0,002%
- LDH ≤0,01%
- Fumarase ≤0,01%
One unit will cause the oxidation of 1,0 µmole of NADH per minute at pH 7,4 and 25 °C.
Malate dehydrogenase is an enzyme that reversibly catalyses the oxidation of malate to oxaloacetate using the reduction of NAD+ to NADH.
Malate dehydrogenase can be used for enzymatic determination of L-malate and gluamate oxaloacetate transaminase (GOT).
- Form:Powder
- Enzyme name:Malate dehrdrogenase
Frequently Bought Together