Collagenase, MP Biomedicals
Catalog # ICNA0215070401
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:1 g
- Enzyme name:Collagenase
Specifications
About this item
For typical assay, dissolve enzyme at a concentration of 1 mg/ml in 0,05 M TES with 0,36 mM calcium chloride, pH 7,5. Dilutions run are 1/10 and 1/20 in the above buffer.
One unit liberates one µmole of L-leucine equivalents from collagen in 5 hours at 37 °C, pH 7,5.
Activators: Ca²⁺ is required; Inhibitors: Metal chelating agents such as cysteine, EDTA or o-phenanthroline but not DFP. It is also inhibited by a2-macroglobulin.