Pyruvate kinase, MP Biomedicals
Dissolves readily at 5 mg/ml in 0,06 M Tris/HCI pH 7,4 to give a clear colorless solution.
- Protein determined by biuret
That amount of enzyme causing the oxidation of one micromole of NADH per minute at 25 °C and pH 7,4.
- CAS No.:9001-59-6
- Enzyme name:Pyruvate kinase
Frequently Bought Together