
Ammonium chloride, powder ACS
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:12125-02-9
:
Formula:NH₄Cl
MDL Number:MFCD00011420
:53.49 g/mol
Melting Pt:335 °C (dec., lit.)
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Assay:≥99,5%
- pH (5% aq soln):4,5 - 5,5
- Insoluble Matter:≤0,005%
- Residue on Ignition:≤0,01%
- Calcium:≤0,001%
- Magnesium:<5 ppm
- Heavy Metals:≤5 ppm
- Iron:<2 ppm
- Phosphate:<2 ppm
- Sulphate:<0,002%
Specification Test Results
Frequently Bought Together