Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Polysorbate 20, liquid

Polysorbate 20, liquid

Supplier:   MP Biomedicals
Polysorbate 20, liquid
Supplier:  MP Biomedicals
Certificates
Polysorbate 20, liquid

3 Options Available Below

Product Details & Documents

Polysorbate 20 is nonionic surfactant derived from sorbitan ester.

  • Refractive Index: n20/D 1.468(Lit.)
  • Viscosity: 400 cps at 25 °C.(Lit.)
  • Critical Micelle Concentration (CMC): 0.06mM

Polysorbate 20 is typically used as an emulsifying agent for the preparation of stable oil-in-water emulsions, particularly in pharmaceutical applications. It has been used in pre-extraction of membranes to remove peripheral proteins. It has been used as a blocking agent on nitrocellulose.

Caution: Research Use Only (RUO).

Synonyms:  Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
  • CAS Number:9005-64-5
  • Formula:C₅₈H₁₁₄O₂₆
  • MDL Number:MFCD00165986
  • Boiling Pt:>100 °C (lit.)
  • Density:1.095 g/mL (lit.)
  • Storage Temperature:Ambient
Specifications for Product Family

Specification Test Results

  • Identity Test A:
    Passes
  • Identity Test B:
    Passes
  • FTIR:
    Conforms to Reference
  • Solubility (1g/20mL):
    Clear, Foamy Solution
  • Specific Gravity:
    1.08-1.113
  • Water Content:
    ≤3%
  • Saponification Number:
    40-50
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 100 mL
    Log in for availability
    Log in for availability
    1 * 100 mL
    1 * 100 mL - SFr. 41.40
    SFr. 41.40
    1 * 100 mL
       SFr. 41.40

  • 500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - SFr. 87.90
    SFr. 87.90
    1 * 500 mL
       SFr. 87.90

  • 1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - SFr. 131.00
    SFr. 131.00
    1 * 1 L
       SFr. 131.00

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..