Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Phenol, white solid, Ultrapure

Phenol, white solid, Ultrapure

Bottle, 500 g
Catalog # ICNA04818048
Corrosive
Toxic
Health
Supplier:   MP Biomedicals
Phenol, white solid, Ultrapure
Catalog # ICNA04818048
Supplier:  MP Biomedicals
CAS Number:  108-95-2
Phenol, white solid, Ultrapure

Bottle, 500 g

CAS Number:  108-95-2
Price
SFr. 318.00
Log in for availability
1 * 500 g
Product Details & Documents

Phenol is useful for extraction of viral, plasmid and phage DNAs, and to solubilise and dissociate proteins. It is used as a general disinfectant, either in solution or mixed with slaked lime, etc., for toilets, stables, cesspools, floors, drains, etc.; for the manufacture of colorless or light-colored artificial resins, many medical and industrial organic compounds and dyes; as a reagent in chemical analysis. Aqueous solutions are used as topical anesthetic; topical antiseptic; topical antipruritic. Phenol is used in DNA extractions for removing protein.

  • Soluble in aqueous solution (5% - clear, colorless solution)

Note: Phenol is prone to redden on exposure to air and light, hastened by presence of alkalinity. This oxidation makes it unsuitable for nucleic acid work. Oxidation products make DNA resistant to hydrolysis by DNase I. One such oxidation product is o-hydroxybiphenyl. Inhibition is due to the direct action on the DNA rather than on the enzyme, due to hydrogen bonding between bases of the nucleic acid.

Soluble in aqueous solution (5% - clear, colorless solution)

Synonyms:  Phenylalcohol, Carbolic acid, Hydroxybenzene
  • CAS Number:108-95-2
  • Formula:C₆H₅OH
  • MDL Number:MFCD00002143
  • Molecular Weight:94.11 g/mol
  • Storage Temperature:Ambient
  • UN:1671
  • ADR:6.1,II
Specifications

Specification Test Results

  • Identity Test:
    Passes
  • Melting Point:
    37 - 44 °C
  • Moisture Content:
    <0,5%
  • Heavy Metals:
    <0,01%

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.