Order Entry
Switzerland
ContactUsLinkComponent
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99%, white crystalline powder, Ultrapure
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99%, white crystalline powder, Ultrapure
Supplier:  MP Biomedicals
Certificates
undefined
TRIS HCl (tris(hydroxymethyl)aminomethane hydrochloride) ≥99%, white crystalline powder, Ultrapure
Supplier:  MP Biomedicals

Specification Test Results

  • FTIR:
    Conforms to Standard
  • pH (5% aq soln) :
    4.0 - 5.0
  • Melting Point :
    145 - 158°C
  • OD260 nm (5% aq soln):
    ≤0.1
  • OD280 nm (5% aq soln):
    ≤0.1
  • Heavy Metals:
    <5 ppm
  • Purity:
    ≥99%

Specification Test Results

Frequently Bought Together