Order Entry
Switzerland
ContactUsLinkComponent
Human ADAM12 ELISA kit
Human ADAM12 ELISA kit
Catalog # BIRBORB99234-96
Supplier:  Biorbyt
CAS Number:  
undefined
Human ADAM12 ELISA kit
Catalog # BIRBORB99234-96
Supplier:  Biorbyt
CAS Number:  

Specifications

  • Pk:
    96
  • Primary antibody reactivity:
    Human
  • Description:
    Human ADAM12 ELISA kit, 96-well
  • Detection range:
    156 - 10000 pg/ml
  • Sensitivity:
    <10 pg/ml

Specifications

About this item

The Human ADAM12 ELISA Kit is used for quantitative detection of human ADAM12 in cell culture supernates, serum, plasma (heparin) and urine.

  • There is no detectable cross-reactivity with other relevant proteins

ADAM12 (disintegrin and metalloproteinase domain-containing protein 12), also known as MLTN, is an enzyme that in humans is encoded by the ADAM12 gene.