Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Mouse decorin ELISA kit

Mouse Decorin ELISA Kit, 96 Tests
Catalog # BIRBORB96784-96
Marketsource
Supplier:   Biorbyt
Mouse decorin ELISA kit
Catalog # BIRBORB96784-96
Supplier:  Biorbyt
Marketsource
CAS Number:  
Mouse decorin ELISA kit
Mouse decorin ELISA kit

Mouse Decorin ELISA Kit, 96 Tests

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 96 Tests
Product Details & Documents

Mouse Decorin ELISA Kit is a sandwich ELISA kit for the quantitative detection of mouse decorin.

  • No detectable cross-reactivity with other relevant proteins

Decorin is a proteoglycan on average 90 to 140 kilodaltons kD in size. It belongs to the small leucine-rich proteoglycan SLRP family and consists of a protein core containing leucine repeats with a glycosaminoglycan GAG chain consisting of either chondroitin sulfate CS or dermatan sulphate DS. Using a genomic clone as the labeled probe and in situ hybridisation of human metaphase chromosomes, decorin gene was mapped to the discrete region of human chromosome 12q23. Decorin is capable of suppressing the growth of various tumour cell lines when expressed ectopically.

 

Specifications

Specifications

  • Pk:
    96 Tests
  • Description:
    Mouse Decorin ELISA Kit
  • Cat. No.:
    BIRBORB96784-96
  • Specificity:
    Natural and recombinant mouse Decorin
  • Sensitivity:
    <10 pg/ml
  • Primary antibody reactivity:
    Mouse
  • Detection range:
    31,2 - 2000 pg/ml

Specifications