Order Entry
Switzerland
ContactUsLinkComponent
Fmoc-Dpr(Boc-Aoa)-OH ≥96.0% (by HPLC) (99.5% ee), Novabiochem®
Zertifikate
Fmoc-Dpr(Boc-Aoa)-OH ≥96.0% (by HPLC) (99.5% ee), Novabiochem®

Ergebnisse des Spezifikationstests

  • Alternate Names:
    N-α-Fmoc-N-β-(N-t.-Boc-amino-oxyacetyl)-L-diaminopropionic acid
  • Colour (Visual):
    White to Slight Yellow to Beige
  • Appearance of Substance (Visual):
    Powder
  • Identity (IR):
    Passes Test
  • Enantiomeric Purity:
    ≥ 99,5% (a/a)
  • Purity (TLC(CMA2)):
    ≥ 98%
  • Assay (HPLC, area%):
    ≥ 96,0% (a/a)
  • Solubility (1 mmole in 2 ml DMF):
    clearly soluble
  • Assay (Acidimetric):
    ≥ 95,0%
  • Water (K. F.):
    ≤ 2,00%
  • Storage:
    Store below +30 °C

Ergebnisse des Spezifikationstests

Frequently Bought Together