Potassium dihydrogen phosphate, anhydrous ≥99%, white crystalline powder ACS
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:7778-77-0
:
Formula:KH₂PO₄
MDL Number:MFCD00011401
:136.09 g/mol
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Assay:≥99%
- pH (5% aq soln, 25 °C ):4,1 - 4,5
- Insoluble Matter:≤0,01%
- Loss on Drying (105 °C):≤0,2%
- Chloride (Cl):≤0,001%
- Sulphate (SO₄):≤0,003%
- Heavy Metals (as Pb):<0,001%
- Iron (Fe):≤0,002%
- Sodium (Na):≤0,005%
Specification Test Results
Frequently Bought Together