Ammoniumsulfat ≥99%, Pulver ACS
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:7783-20-2
:
Formula:(NH₄)₂SO₄
MDL Number:MFCD00003391
:132.14 g/mol
Storage Temperature:Raumtemperatur
Specification Test Results
- Assay:≥99%
- pH (5% aq soln):5 - 6
- Insoluble Matter:≤0.005%
- Residue on Ignition:≤0,005%
- Chloride (Cl):≤5 ppm
- Nitrate (NO₃):≤0,001%
- Phosphate (PO4):≤5 ppm
- Heavy Metals (as Pb):≤5 ppm
- Iron (Fe):≤5 ppm
Specification Test Results
Frequently Bought Together