Order Entry
Switzerland
ContactUsLinkComponent
Amylase, MP Biomedicals
Amylase, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Amylase, MP Biomedicals
Supplier:  MP Biomedicals
Certificates

Specifications

  • CAS No.:
    9000-92-4
  • Form:
    Powder
  • Enzyme name:
    Amylase

Specifications

Related Information

Frequently Bought Together