Kryoboxen B55/B75/B95-Serien
Anbieter: GLW
New Product
Zertifikate
Über diesen Artikel
Cryoboxes made of PP, natural or coloured.
- Autoclavable up to +121 °C and use in freezer down to –90 °C
- Included clear lid offers a view of the contents without opening the cryo-box
- Drainage holes in base for easy opening the box at low temperatures
- For light sensitive samples please choose the colour Black/black
Suitable for 25 vials, arranged in 5×5 compartments with 21,6 mm diameter. Suitable to store up to 25 HPLC vials, GC vials, tubes or wide-neck bottles and other flat-bottomed vials fit in the grid. The numeric labeling helps to identify your samples.
B55-Series:
Generic tube size up to 53 mm height and 21,6 mm diameter.
B75-Series:
Generic tube size up to 78 mm height and 21,6 mm diameter.
B95-Series:
Generic tube size up to 93 mm height and 21,6 mm diameter.
Frequently Bought Together