ContactUsLinkComponent
Water treatment units, Aqua purificator PG 8595
Water treatment units, Aqua purificator PG 8595
Fournisseur:  MIELE
Certificats
undefined
Water treatment units, Aqua purificator PG 8595
Fournisseur:  MIELE
Spécifications
  • Catalog No:
  • MIEL10087320
  • MIEL10087310
  • Model:
  • PG 8595
  • PG 8595
  • Water quality:
  • <19 µS/cm
  • <19 µS/cm
  • W×D×H:
  • 300×600×835 mm
  • 300×600×835 mm
  • Weight:
  • 26 kg
  • 26 kg
Spécifications
Frequently Bought Together