Order Entry
Switzerland
ContactUsLinkComponent
Methanol ≥99.9%, HiPerSolv CHROMANORM® ACS, Reag. Ph. Eur., super gradient grade for HPLC, suitable for UPLC/UHPLC instruments
Flammable
Toxic
Health
Methanol ≥99.9%, HiPerSolv CHROMANORM® ACS, Reag. Ph. Eur., super gradient grade for HPLC, suitable for UPLC/UHPLC instruments
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Flammable
Toxic
Health
Methanol ≥99.9%, HiPerSolv CHROMANORM® ACS, Reag. Ph. Eur., super gradient grade for HPLC, suitable for UPLC/UHPLC instruments
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Assay (GC):
    Min. 99.9 %
  • Carbonyl compounds:
    Passes test
  • Identification:
    Passes test
  • Solubility in water:
    Passes test
  • Substances darkened by sulphuric acid:
    Passes test
  • Substances reducing permanganate:
    Passes test
  • Acidity:
    Max. 0.0002 meq/g
  • Alkalinity:
    Max. 0.0002 meq/g
  • Boiling point:
    64 - 65 °C
  • Colouration:
    Max. 10 APHA
  • Density (20/20):
    0.791 - 0.793
  • Evaporation residue:
    Max. 1 ppm
  • Water:
    Max. 0.02 %
  • Gradient (235 nm):
    Max. 2 mAU
  • Gradient (254 nm):
    Max. 1 mAU
  • Transmittance (210 nm):
    Min. 45 %
  • Transmittance (220 nm):
    Min. 65 %
  • Transmittance (225 nm):
    Min. 70 %
  • Transmittance (230 nm):
    Min. 83 %
  • Transmittance (235 nm):
    Min. 85 %
  • Transmittance (240 nm):
    Min. 90 %
  • Transmittance (250 nm):
    Min. 95 %
  • Transmittance (260 nm):
    Min. 98 %
  • Transmittance (280-400 nm):
    Min. 98 %
  • Absorbance (210 nm):
    Max. 0.347
  • Absorbance (220 nm):
    Max. 0.188
  • Absorbance (225 nm):
    Max. 0.155
  • Absorbance (230 nm):
    Max. 0.081
  • Absorbance (235 nm):
    Max. 0.071
  • Absorbance (240 nm):
    Max. 0.046
  • Absorbance (250 nm):
    Max. 0.023
  • Absorbance (260 nm):
    Max. 0.009
  • Absorbance (280-400 nm):
    Max. 0.009
  • Fluorescence (as quinine; 254 nm):
    Max. 1 ppb
  • Fluorescence (as quinine) (365 nm):
    Max. 0.5 ppb
  • Conforms Ph.Eur. R1 1053201:
    Passes test
  • Conforms Ph.Eur. R2 1053202:
    Passes test
  • Conforms to ACS:
    Passes test
  • Filtered at 02 µm:
    Confirmed

Specification Test Results

Frequently Bought Together