Rust Grade Book, ISO 8501-1
Illustrated rust grade book
Catalog # SIMELD3020
Supplier: Industrial Physics
Jump to:
Product Details & Documents
Pictorial standard book with rust grades and preparation grades of uncoated steel substrates and of steel substrates after overall removal of previous coatings. Also called 'Swedish Blast Degrees' or Sa-standards). The original visual standard. It shows the degree of cleanliness of four different levels of rusted steel cleaned by grit blasting, hand/power tool and flame cleaning.
Documents
TQC Company Video
Videos
Specifications
- Description:Illustrated rust grade book