Reference manual for coating defects, Fitz's Atlas 2
N° de catalogue SIMELD3061
Fournisseur: Industrial Physics
Spécifications
- Description:Fitz's Atlas 2 for coating defects, English only
- Pk:1
Spécifications
À propos de cet article
Fitz's Atlas 2 is a reference manual of colour images with a concise description of every problem and their associate causes.
The book is divided in sections such as weld faults, presurface conditions, coating defects, microscopy and marine fouling.