Order Entry
Switzerland
ContactUsLinkComponent
Disposable drain seal
Disposable drain seal
Catalog # NEWPPLRE166
Supplier:  NEW PIG
undefined
Disposable drain seal
Catalog # NEWPPLRE166
Supplier:  NEW PIG

Specifications

  • Description:
    Disposable drain seal, grey
  • L×W:
    650×450 mm
  • Pk:
    4

Specifications

About this item

Versatile product stops liquids from entering drains and causing pollution. Clay composition creates a 100 % seal, even sticking to rough, irregular surfaces like tarmac or concrete.

  • Easy to use - Simply peel of the protective plastic from one side, then press the exposed side firmly into and around the drain
  • Resistant to the most common oils and chemicals
  • Flexible at low temperatures to –20 °C and treated with a formula which prevents mould growth
Certifications: Help with ADR compliance when a drain seal is required.