ContactUsLinkComponent
No Cal stain lifter solution, Orion™
No Cal stain lifter solution, Orion™
Catalog # ORIO510015
Supplier:  Thermo Orion
No Cal stain lifter solution, Orion™
Catalog # ORIO510015
Supplier:  Thermo Orion
Specifications
  • Description:
    No Cal stain lifter solution, Orion™, 60 ml bottle
  • Pk:
    1
Specifications
About this item
This cleaning solution restores proper electrode performance and prolongs the useful life of the electrode.