Order Entry
Switzerland
ContactUsLinkComponent
Medical absorbent mats
Medical absorbent mats
Catalog # NEWPMATE1110
Supplier:  NEW PIG
undefined
Medical absorbent mats
Catalog # NEWPMATE1110
Supplier:  NEW PIG

Specifications

  • Description:
    Absorbent mat roll, lightweight, thickness = 1,3 mm
  • L×W:
    50 m×900 mm
  • Seq:
    01
  • Absorbency:
    72 l/bag
  • Pk:
    1

Specifications

About this item

These medical absobent mats are made from a non-wetting top layer to keep the standing surface dry, a super-absorbent central layer and a non-slip backing that grips to the floor. The mats keep floors dry during wet operating procedures.

  • Reduces slips and falls
  • Easily lifted and disposed of after use which reduces clean up time
  • Minimises tripping hazards
  • Reduces clean up time, it can be easily lifted and disposed after use
  • Simple to use, can be cut into individual sizes

For hospital floors, it absorbs spills from procedures like knee and hip operations where saline fluids are used in abundance for irrigation.

Composed of three layers: The top layer is three-dimensional perforated LDPE; the core is made from air-laid cellulose with PP; the base layer is a PE high-grip film.