ContactUsLinkComponent
Acrylamide/bis premix, 29:1, MP Biomedicals
Acrylamide/bis premix, 29:1, MP Biomedicals
Catalog # ICNA04800657
Supplier:  MP Biomedicals
Acrylamide/bis premix, 29:1, MP Biomedicals
Catalog # ICNA04800657
Supplier:  MP Biomedicals
Specifications
  • Pk:
    30 g
Specifications
About this item
Premix of ultra pure acrylamide with the cross-linking reagent N,N'-methylene-bis-acrylamide, final ratio 29:1.