Order Entry
Switzerland
ContactUsLinkComponent
Duffle Bag, Lockout
Duffle Bag, Lockout
Catalog # BRDY099162
Supplier:  Brady
undefined
Duffle Bag, Lockout
Catalog # BRDY099162
Supplier:  Brady

Specifications

  • Colour:
    Blue/black
  • Description:
    Dufflebag, with shoulder strap (95 mm) and pockets (8)
  • W×D×H:
    420×125×215 mm
  • Pk:
    1

Specifications

About this item

These duffle bags are made of 600 denier polyester that are used for lock out kits.