ContactUsLinkComponent
Culture tubes, screw thread
Culture tubes, screw thread
Catalog # SCERD61251600
Supplier:  DWK Life Sciences
Culture tubes, screw thread
Catalog # SCERD61251600
Supplier:  DWK Life Sciences
Specifications
  • Ø:
    16 mm
  • Height:
    125 mm
  • Pk:
    100
Specifications
About this item
Tubes, made up of borosilicate glass, grade 3.3, with screw threads.
  • Caps must be ordered separately
  • White marking spot
  • Very good chemical and temperature resistance