Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Electrode cleaning solution, RENOVO.X

Electrode cleaning solution, RENOVO.X

Extra strong electrode cleaning solution, RENOVO.X, 250 ml bottle, 250 mL
Catalog # HACHS16M002
Supplier:   Hach
Electrode cleaning solution, RENOVO.X
Catalog # HACHS16M002
Supplier:  Hach
CAS Number:  
Electrode cleaning solution, RENOVO.X
Electrode cleaning solution, RENOVO.X

Extra strong electrode cleaning solution, RENOVO.X, 250 ml bottle, 250 mL

CAS Number:  
Price
43.40 SFr.
Log in for availability
1 * 250 mL
Product Details & Documents

This extra strong electrode cleaning solution is ideal for use when stronger cleaning agents are required, for example when the solutions contain protein.

Documents

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Extra strong electrode cleaning solution, RENOVO.X, 250 ml bottle

Specifications