Cleanroom Overalls Tyvek® IsoClean® model IC183B DS
Anbieter: DuPont
Zertifikate
Über diesen Artikel
These unhooded overalls are made of Tyvek®, a fabric composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µm in size).
- Ultra-low linting fabric
- Unhooded overall with bound neck
- Tyvek® covered elasticated thumb loops
- Tunnelled elastication at wrists and ankles; stitched-in elasticated waist
- Front zipper closure with storm flap
- Optimised shape and fit allowing full freedom of movement
- Clean-processed and gamma-sterilised
- Gamma irradiated to Sterility Assurance Level (SAL) of 10⁻⁶
Garments made of Tyvek® IsoClean are used in the biotech, pharmaceutical, medical device manufacturing and electronics industries, as well as in other critical or controlled environments.
Colour: White
Certifications: Type 5B/6B, EN 1073-2 (Class 2), EN 14126, Cat. III
Packaging: Double bagged. Packed in an ISO Class 4 certified cleanroom .
Technische Daten
- Catalog No:
- 113-8629
- 113-8628
- 113-8633
- 113-8632
- 113-8631
- 113-8630
- DUPOIC183BWHDS/XS
- Model:
- IC 183 B
- IC 183 B
- IC 183 B
- IC 183 B
- IC 183 B
- IC 183 B
- IC 183 B
- Garment application:
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical
- Chemical protection, Cleanroom, Medical, Laboratory, Clinical
Technische Daten
Zugehörige Informationen
Cleanroom_Whitepaper_Advantages of IsoClean_EN_Final DUPONT™ TYVEK® ISOCLEAN®Sizing Chart model IC 183 B DS
Anwendungshinweise
Controlled_Environments_Catalogue_2016_EN DUPONT™ TYVEK® ISOCLEAN® DUPONT™ TYVEK® ISOCLEAN® FAQ Poster Non-Sterile Cleanroom Gowning Procedure
Produktliteratur
DUPOIC183BWHDS^XL_TDS_EN
Technisches Datenblatt
Frequently Bought Together