Orders LinkContactUsLinkComponent
Baumé hydrometers with thermometer
Baumé hydrometers with thermometer
Supplier:   Amarell
Baumé hydrometers with thermometer
Supplier:  Amarell
Certificates
Baumé hydrometers with thermometer
4 Options Available Below
Product Details & Documents
Baumé hydrometers with thermometer calibrated at 15 °C.
  • Red or blue filling liquid in round capillary
  • Paper scale thermometers designed with well visible print
  • Shot weighted body at the bottom pointed end
Thermometer is presented inside the hydrometer body allowing for simultaneous temperature measurements.
Product Family Options
Product Information
  • Range
    Division
    Temperature Range
    Length
    Availability
    Price
  • AMARH843210-FL
    Baumé hydrometer with thermometer
    Baumé hydrometer with thermometer
    0 - 30 °Bé
    1,0 °Bé
    0…40 °C
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 88.80
    SFr. 88.80
    1 * 1 ST
       SFr. 88.80
  • AMARH843288-FL
    Baumé hydrometer with thermometer
    Baumé hydrometer with thermometer
    0 - 50 °Bé
    1,0 °Bé
    0…40 °C
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 88.80
    SFr. 88.80
    1 * 1 ST
       SFr. 88.80
  • AMARH843290-FL
    Baumé hydrometer with thermometer
    Baumé hydrometer with thermometer
    0 - 70 °Bé
    1,0 °Bé
    0…40 °C
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 88.80
    SFr. 88.80
    1 * 1 ST
       SFr. 88.80
  • AMARH843216-FL
    Baumé hydrometer with thermometer
    Baumé hydrometer with thermometer
    30 - 70 °Bé
    1,0 °Bé
    0…40 °C
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 88.80
    SFr. 88.80
    1 * 1 ST
       SFr. 88.80