ContactUsLinkComponent
ADC active digital controllers
ADC active digital controllers
Catalog # EDWAD39591500
ADC active digital controllers
Catalog # EDWAD39591500
Specifications
  • Accuracy:
    1×10ˉ9 mbar
  • Description:
    Active digital controllers, ADC mkII Enhanced
  • Model:
    ADC mkII Enhanced
  • Permitted ambient temperature for operation:
    +0...+40 °C
  • Permitted ambient temperature for storage:
    −30...+70 °C
  • Pk:
    1
Specifications
About this item
This Active Digital Controller (ADC) is a compact single gauge controller with automatic recognizes compatible Edwards gauges, loads the appropriate look-up table and displays the pressure in commonly used vacuum units.
  • Bright LED display gives clear, long distance readability
  • The ADC displays units in mbar, Torr, Pascal or Volts
  • Simple push-button control makes operation highly intuitive
  • This gauges are mainly used in laboratory and industrial applications
The enhanced controller supports two similar gauges - it has two variable hysteresis set-points which are linked to 48 V DC. 1 A changeover relays and two 0 to 10 V DC analog outputs. To aid system integration, the enhanced controller is provided with an RS232 interface. When combined with a suitable gauge, such as the Edwards APGXH Convection Pirani or Wide Range Gauge (WRG), the ADC represents a cost effective means of monitoring and controlling process vacuum in a broad range of applications.