ContactUsLinkComponent
Particulate filter, PlexTec P3
Particulate filter, PlexTec P3
Catalog # 111-1916
Supplier:  M S A
undefined
Particulate filter, PlexTec P3
Catalog # 111-1916
Supplier:  M S A
Specifications
  • Type:
    PlexTec P3 particle filter with round thread
  • Pk:
    1
  • Class:
    P3 R
Specifications
About this item
These particle, gas and combined filters are suitable for the Advantage 410 half mask and for Advantage 3100, Ultra Elite, 3S Basis Plus and 3S full masks. With round thread (EN 148-1).