ContactUsLinkComponent
Universal body, Omni-Cell™
Universal body, Omni-Cell™
Catalog # SPCC1800
Supplier:  Specac
Universal body, Omni-Cell™
Catalog # SPCC1800
Supplier:  Specac
Specifications
  • Description:
    Universal body, Omni-Cell™
  • Pk:
    1
Specifications
About this item
Omni-Cell is an approach to the analysis of liquid samples in transmission spectroscopy – One cell is suitable for all applications.
  • Quick to assemble and change windows
  • Windows and spacers compatible with older cells compatible for FTIR and dispersive spectroscopy
  • Quick release clamping mechanism, wide choice of window materials
Compatible with all FTIR spectrometers as well as older dispersive systems. Can be configured for use as demountable liquid cells permanently sealed liquid cells or as mull cells.
Delivery: Includes front and back plates, cell nest, quick release nuts (4), bonded front PTFE gasket, rear neoprene gasket and PTFE Luer plugs (2).