ContactUsLinkComponent
Anion exchange columns, Dionex™, IonPac™ AS18
 :  DIONEX
Anion exchange columns, Dionex™, IonPac™ AS18
 :  DIONEX
  • Catalog No:
  • DION060549
  • DION060551
  • DION060553
  • DION060555
  • Max. pressure:
  • 4000 psi
  • 4000 psi
  • 4000 psi
  • 4000 psi
  • pH range:
  • 0 - 14
  • 0 - 14
  • 0 - 14
  • 0 - 14
Frequently Bought Together