ContactUsLinkComponent
Water purification system, WaterPro® BT
Water purification system, WaterPro® BT
Supplier:  LABCONCO
Certificates
undefined
Water purification system, WaterPro® BT
Supplier:  LABCONCO
Specifications
  • Catalog No:
  • LBCP9015030
  • LBCP9015130
  • Model:
  • WaterPro BT
  • WaterPro BT (with UV lamp)
  • Feed water:
  • Tap
  • Tap
  • Flow rate:
  • >0,5 l/min
  • >0,5 l/min
  • TOC content:
  • <10 ppb
  • <5 ppb
  • W×D×H:
  • 290×422×538 mm
  • 290×422×538 mm
  • Weight:
  • 17,6 kg
  • 18,2 kg