Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Agar, Bacteriological Grade

Agar, Bacteriological Grade

Supplier:   G-Biosciences
Agar, Bacteriological Grade
Supplier:  G-Biosciences
Certificates
Agar, Bacteriological Grade
Agar, Bacteriological Grade

2 Options Available Below

Product Details & Documents

A purified agar that is a fine light colored powder that is used as a solidification agent in the preparation of microbiological culture media.

Bacteriological grade purity: Low contaminant levels suitable for sensitive microbiological applications.

Synonyms:  Agar Agar
  • CAS Number:9002-18-0
  • Formula:(C₁₂H₁₈O₉)n
  • MDL Number:MFCD00081288
  • Melting Pt:90 °C
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - SFr. 227.00
    SFr. 227.00
    1 * 500 g
       SFr. 227.00

  • Bottle
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - SFr. 430.00
    SFr. 430.00
    1 * 1 kg
       SFr. 430.00