Safety helmets, uvex pheos B-S-WR
Supplier: UVEX
Certificates
About this item
Lightweight helmets made from HDPE.
- Three variable air vents aids airflow and helps regulate the temperature within the helmet making it more comfortable to wear
- Adapter systems for attaching uvex visors, uvex earmuffs and lamps
- Suspension harness with rotating ratchet wheel allows the width size to be adjusted for an individual fit
- Provides protection even in extremely low temperatures
Adjustable size range: 520 to 610 mm
Certifications: EN 397 with the additional requirement: –30 °C; MM = molten metal (splash)
Frequently Bought Together