Cell counter, 'easy read'
Codice catalogo TPPA87010
Nome del fornitore: TPP
Specifiche
- Range:0 - 5 µl
- Description:Easy read measuring device for read-out process with PCV tubes
- Material:Aluminium
- W×D×H:320×33×38 mm
- Pk:1
Specifiche
Informazioni su questo articolo
These 'easy read' measuring device are fast, easy to use, reproducible and accurate alternative to commonly used methods for determination of a cell number density. The read-out process of the cell mass in the PCV tube with this requires neither special training nor sophisticated expensive machines.
- Accurate and reproducible cell pellet measuring
- Solid re-usable and easy handling measuring device
- Read-out device with magnifying glass and cross line
Optimal with the use of PCV tubes.