Verification standard kit, MultiDirect
Supplier: Lovibond
Jump to:
About this item
Verification accuracy of photometers and validity of results.
- Check the performance of photometers over the full measurement range
- Results can be checked against tolerances provided on the certificate of conformity
: Contains zero solution in sealed photometer vial, 6× verification standard solution in sealed photometer vial, light shield cap, certificate of conformity, instruction sheet.
2 Options Available Below
Product Details & Documents
Verification accuracy of photometers and validity of results.
- Check the performance of photometers over the full measurement range
- Results can be checked against tolerances provided on the certificate of conformity
: Contains zero solution in sealed photometer vial, 6× verification standard solution in sealed photometer vial, light shield cap, certificate of conformity, instruction sheet.
Product Family Options
Product Information
- PkAvailabilityPrice
- LOVI215650Verification standard kit, Multidirect 0/430/530/560/580/610/660 nmVerification standard kit, Multidirect 0/430/530/560/580/610/660 nm1 KITLog in for availability1 * 1 KITSFr. 315.00
- LOVI215670Verification standard kit, MD 100/MD200