ContactUsLinkComponent
Racks for microtubes and cryo vials
Racks for microtubes and cryo vials
Catalog # KOEH2-1642
Racks for microtubes and cryo vials
Catalog # KOEH2-1642
Specifications
  • No. of Holes:
    8x8
  • Colour:
    Red/white
  • Description:
    Compartment size: 12,7x12,7 mm
  • W×D×H:
    143×143×34 mm
  • Pk:
    1
Specifications
About this item
PP rack, square, easy to mount. For 1,5/2,0 ml mircrocentrifuge tubes and cryogenic vials.
  • Alphanumeric index of the holes
  • Solid bottom of each well to secure vials in place for easier opening and closing
  • Fits perfectly and is stackable even when filled with tubes