ContactUsLinkComponent
Condensers, Allihn, with Quickfit® ground joints
Condensers, Allihn, with Quickfit® ground joints
Catalog # 201-8312
Supplier:  SciLabware
undefined
Condensers, Allihn, with Quickfit® ground joints
Catalog # 201-8312
Supplier:  SciLabware
Specifications
  • Cone size:
    24/29* NS
  • Jacket length:
    400 mm
  • Socket size:
    24/29 NS
  • Seq:
    07
  • Pk:
    1
Specifications
About this item
Condensers acc. to Allihn, PYREX® borosilicate glass, for condensing vapours.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for reflux applications. With NS socket and NS cone. Inner bulb sections provide increased cooling surface. With detachable plastic screwthread connectors for easy and safe attachment of flexible tubing.