Turbidity standards, mid range 200 to 1000 NTU
Anbieter: REAGECON
Zertifikate
Über diesen Artikel
Turbidity standards for ratio and non-ratio instruments composed of suspended polymer microspheres.
- Non toxic and non carcinogenic microspheres
- Highly accurate with well defined spherical shape
- Traceable to NIST
- US EPA approved
- Ready to use- particles stay in suspension
Frequently Bought Together