ContactUsLinkComponent
Syringe priming kit
Syringe priming kit
Catalog # HAMIPRMKIT
Supplier:  HAMILTON BONADUZ
undefined
Syringe priming kit
Catalog # HAMIPRMKIT
Supplier:  HAMILTON BONADUZ
Specifications
  • Description:
    Priming kit
  • Pk:
    1 KIT
Specifications
About this item
The priming kit provides the necessary tools to ensure that the syringe and needle connection is completely primed. The glass capillary tubing traps a large volume of liquid that can only be overcome by using the priming kit. The kit is compatible with 600, 700, 1700, removable needle syringes and 7000 series syringes.
Delivery information: Kit includes one 250 µl syringe, one 6 pack of 30 gauge needles and one pack of septa.