0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Forceps
Artery forceps, Krönlein type, Aesculap®
Artery forceps, Krönlein type, Aesculap®
Artery forceps, Krönlein type, Aesculap®
Catalog # AESCBH300R
Supplier:
Aesculap
Expand image
Artery forceps, Krönlein type, Aesculap®
Catalog # AESCBH300R
Supplier:
Aesculap
Specifications
Type:
Krönlein, straight, blunt
Length:
130 mm
Seq:
03
Pk:
1
Specifications
About this item
Product Details
Product Details